Anti-TTR Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-0152R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Transthyretin
- Antigen symbol:TTR
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:22139
- Antigen synonyms:HEL111|CTS1|TBPA|HsT2651|Transthyretin|CTS|PALB|ATTR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:45-95/147
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Thyroid hormone-binding protein. Probably transports thyroxine from the bloodstream to the brain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TTR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat