Order Entry
Denmark
ContactUsLinkComponent
Anti-CD3-delta Rabbit Polyclonal Antibody
Anti-CD3-delta Rabbit Polyclonal Antibody
Catalog # BWRLBS6280
Anti-CD3-delta Rabbit Polyclonal Antibody
Catalog # BWRLBS6280

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD3- delta
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant full length Human CD3-delta.

CD3-? pAb detects endogenous levels of CD3-? protein.

Type: Primary
Antigen: CD3-delta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human