Anti-Ang-1 Rabbit Polyclonal Antibody [clone: E207]
BWRLBS2829
- Antibody type:Primary
- Antigen symbol:Ang-1
- Clonality:Polyclonal
- Clone:E207
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 176-224 of Human Ang-1.
Ang-1 (E207) pAb detects endogenous levels of Ang-1 (E207) protein.
Type: Primary
Antigen: Ang-1
Clonality: Polyclonal
Clone: E207
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat