Cleanroom vacuum cleaner, EXP1-10 CR (MRAC)
Catalog # TIGV110343B
Supplier: TIGER-VAC
Specifications
- Airflow:224 m3/h
- Filters:ULPA filter - included with an efficiency of 99,999% on 0,12 micron. Tested: IEST RP-CC001.3. Filtration efficiency of 99,9995% on 0,18 micron. Tested: IEST RP-CC001.3. U15 by MPPS method as per EN 1822.
- Vacuum pressure:1554 mm H2O
- Description:Vacuum cleaner, EXP1-10 CR (MRAC)
- Environmentally Preferable:
- Model:EXP1-10/15 CR (MRAC)
- Horsepower:1,3 hp
- Current:4,5 A
- Frequency:50 Hz
- Power:1000 W
- Voltage:230 V
- Phase:Single
- Pk:1
Specifications
About this item
These wet and dry vacuum cleaner units are cleanroom compatible, autoclavable and can be used in the pharmaceutical industry. These units are also available with a middle ring with activated carbon cartridge for the adsorption of toxic/nuisance fumes and vapors.
- All stainless steel SAE 304 construction
- Ideal for general cleanup in hazardous locations
- Single phase explosion proof/dust ignition proof motor
- Less than 10 ohms of resistivity
- Recovery tank capacity of 38 to 57 litres
- Equipped with a true ULPA Filter (U15) located downstream of the motor
Delivery: Inlcudes complete static dissipating autoclavable tool kit and suction hose.