Chloroquine diphosphate salt
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate
CAS Number:50-63-5
:
Formula:C₁₈H₂₆ClN₃
MDL Number:MFCD00069852
Molecular Weight:515.86 g/mol
Melting Pt:200 °C (dec., lit.)
EINECS:200-055-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Physical Appearance:Off-White Powder
- pH:(1% aq soln) 4.0 - 5.0
- pKa:8.4 and 10.8 (for chloroquine base at 20 °C)
- Solubility:Soluble in water. Practically insoluble in alcohol, benzene, chloroform and ether.
- Typical Working Concentration:Chloroquine showed very high antiviral activity against NiV but very little activity against the other viruses at concentrations lower than 20 µM
Specification Test Results
Frequently Bought Together