Order Entry
Denmark
ContactUsLinkComponent
DNA molecular weight marker, wide range
DNA molecular weight marker, wide range
Catalog # J67051.XI
CAS Number:  
DNA molecular weight marker, wide range
Catalog # J67051.XI
CAS Number:  
Specifications
  • Pk:
    0,5 mL
  • Description:
    DNA MW marker, wide range
  • Environmentally Preferable:
Specifications
About this item
This wide range molecular weight marker contains 60 ng/μl of 21 fragments from 100 bp to 12000 bp. The bands appear in 100 bp increments from 100 to 1000 bp and in 1000 bp increments from 1 to 12 kb. Bands at 1 kb and 5 kb stain brighter to aid identification.