Anti-Ku70 Mouse Monoclonal Antibody [clone: S5C11]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Ku70
Clonality: Monoclonal
Clone: S5C11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB327
- DIABMOB327-05
- Antigen symbol:
- Ku70
- Ku70
- Antigen name:
- 5'-deoxyribose-5-phosphate lyase Ku70
- 5'-deoxyribose-5-phosphate lyase Ku70
- Antigen synonyms:
- CTC box binding factor 75 kDa subunit|DNA repair protein XRCC6|lupus Ku autoantigen protein p70|thyroid|Ku autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|70 kDa subunit|ATP-dependent DNA helicase II|5'-dRP lyase Ku70|5'-deoxyribose-5-phosphate lyase Ku70|CTC box-binding factor 75 kDa subunit|70kDa,X-ray repair cross-complementing protein 6|ATP-dependent DNA helicase 2 subunit 1|thyroid autoantigen 70kD (Ku antigen)|ATP-dependent DNA helicase II 70 kDa subunit
- CTC box binding factor 75 kDa subunit|DNA repair protein XRCC6|lupus Ku autoantigen protein p70|thyroid|Ku autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|70 kDa subunit|ATP-dependent DNA helicase II|5'-dRP lyase Ku70|5'-deoxyribose-5-phosphate lyase Ku70|CTC box-binding factor 75 kDa subunit|70kDa,X-ray repair cross-complementing protein 6|ATP-dependent DNA helicase 2 subunit 1|thyroid autoantigen 70kD (Ku antigen)|ATP-dependent DNA helicase II 70 kDa subunit
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- S5C11
- S5C11
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
Specifications
Frequently Bought Together