Order Entry
Denmark
ContactUsLinkComponent
 
Anti-TMEM16A Mouse Monoclonal Antibody (Biotin) [clone: DOG1.1]
 :  Biotium
 
Anti-TMEM16A Mouse Monoclonal Antibody (Biotin) [clone: DOG1.1]
 :  Biotium

 

  • Catalog No:
  • BNCB0725-500
  • BNCB0725-100
  • Antigen symbol:
  • TMEM16A
  • TMEM16A
  • Antigen name:
  • DOG-1
  • DOG-1
  • Antigen synonyms:
  • ORAOV2|Anoctamin 1|TMEM16A|Discovered On Gastrointestinal Stromal Tumors Protein-1 (DOG-1),TAOS2|Calcium Activated Chloride Channel
  • ORAOV2|Anoctamin 1|TMEM16A|Discovered On Gastrointestinal Stromal Tumors Protein-1 (DOG-1),TAOS2|Calcium Activated Chloride Channel
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone DOG1.1
  • Clone DOG1.1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 503074
  • 503074
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together