Anti-CD38 Mouse Monoclonal Antibody (Purified) [clone: AT1]
Supplier: Biotium
Certificates
About this item
CD38 is a type II transmembrane glycoprotein that is present on early B- and T-cell lineages and activated B- and T-cells but is absent from most mature resting peripheral lymphocytes. CD38 is also found on thymocytes, pre-B cells, germinal center B-cells, mitogen-activated T-cells, monocytes and Ig-secreting plasma cells. CD38 is expressed on CD34 cells. The CD34 CD38- population of hematopoietic stems cells defines the most pluripotent cells (e.g. blast colony forming cells).
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT1
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNUB0430-500
- BNUB0430-100
- Antigen symbol:
- CD38
- CD38
- Antigen synonyms:
- cADP-ribose Hydrolase 1|ADP Ribosyl Cyclase 1|T10|NIM-R5 Antigen|CD38H|Acute Lymphoblastic Leukemia Cells Antigen|p45|NAD( ) Nucleosidase
- cADP-ribose Hydrolase 1|ADP Ribosyl Cyclase 1|T10|NIM-R5 Antigen|CD38H|Acute Lymphoblastic Leukemia Cells Antigen|p45|NAD( ) Nucleosidase
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone AT1
- Clone AT1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 479214
- 479214
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide
Specifications
Frequently Bought Together