Order Entry
Denmark
ContactUsLinkComponent
 
Anti-VCAM1 Mouse Monoclonal Antibody (Biotin) [clone: B-K9]
 :  Biotium
 
Anti-VCAM1 Mouse Monoclonal Antibody (Biotin) [clone: B-K9]
 :  Biotium

 

  • Catalog No:
  • BNCB0304-100
  • BNCB0304-500
  • Antigen symbol:
  • VCAM1
  • VCAM1
  • Antigen name:
  • Vascular cell adhesion molecule 1
  • Vascular cell adhesion molecule 1
  • Antigen synonyms:
  • CD106|Vascular cell adhesion protein 1|INCAM-100|Vascular Cell Adhesion Molecule 1 (VCAM1)|L1CAM
  • CD106|Vascular cell adhesion protein 1|INCAM-100|Vascular Cell Adhesion Molecule 1 (VCAM1)|L1CAM
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone B-K9
  • Clone B-K9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 109225
  • 109225
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together