Order Entry
Denmark
ContactUsLinkComponent
Anti-CKB Mouse Monoclonal Antibody (CF488A) [clone: 2ba6]
Anti-CKB Mouse Monoclonal Antibody (CF488A) [clone: 2ba6]
Supplier:  Biotium
Certificates
Anti-CKB Mouse Monoclonal Antibody (CF488A) [clone: 2ba6]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880196-100
  • BNC880196-500
  • Antigen symbol:
  • CKB
  • CKB
  • Antigen name:
  • Creatine Phosphokinase
  • Creatine Phosphokinase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone 2ba6
  • Clone 2ba6
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 173724
  • 173724
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together