Dehydrated culture media, tryptic soy broth, MP Biomedicals
Catalog # ICNA091010717
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 g
- Standards:-
- Description:Tryptic soya broth
- Environmentally Preferable:
- Sterility:Sterilize by autoclave
- Osmolality:272 mM/kg (Lit.)
- Formulation:Casein Digest Peptone: 17,0 g/L, Soy Peptone: 3,0 g/L, Disodium Phosphate: 2,5 g/L, Dextrose: 2,5 g/L, Sodium Chloride: 5,0 g/L
- pH:7,1 - 7,5
- Physical form:Tan Powder
- Format:Powder
Specifications
About this item
Tryptic Soy Broth is used for the cultivation of fastidious and non-fastidious microorganisms. It is a highly nutritive and versatile medium in the laboratory. This medium promotes abundant growth of fastidious microorganisms without the necessity of adding serum and other materials. It may also be used for the rapid determination of the bacteriological quality of water.
- Tan powder format
- pH 7,1 to 7,5
- Stored at room temperature
- Ideal for bacteria cell culture
Storage temperature : Room temperature (15...30 °C)