Anti-VHL Goat Polyclonal Antibody (Biotinylated)
EVBIEB07333-B
- Antibody type:Primary
- Antigen name:Von Hippel-Lindau Tumor Suppressor
- Antigen symbol:VHL
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:Affinity Purified
- Form:Liquid
- Gene ID:7428
- Antigen synonyms:VHL|elongin binding protein|von Hippel-Lindau tumor suppressor|RCA1|protein G7|pVHL|VHL1|E3 ubiquitin protein ligase|HRCA1
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Molecular weight:24.2; 19.7; 21.3
- Sequence:C-RSLVKPENYRRLD.
- Storage temperature:+8 °C
- Concentration:0,5 µg/µl
- Immunogen:VHL, Biotinylated
- Size:200 μL
- Pack type:Plastic Vial
- Pk:0,1 mg
Type: Primary
Antigen: VHL
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human