Order Entry
Denmark
ContactUsLinkComponent
Anti-IgG Mouse Antibody
Anti-IgG Mouse Antibody
Supplier:  MP Biomedicals
Certificates
Anti-IgG Mouse Antibody
Supplier:  MP Biomedicals

Specifications

  • Catalog No:
  • ICNA08641461
  • ICNA08641462
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Host:
  • Mouse
  • Mouse
  • Form:
  • Lyophilised powder
  • Lyophilised powder
  • Storage temperature:
  • 2...8 °C
  • 2...8 °C
  • Tested applications:
  • Immunoelectrophoresis
  • Immunoelectrophoresis

Specifications

Frequently Bought Together