Dextri-maltose®, coarse powder
Catalog # ICNA0296004801
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Reference
- pH (20% aq soln) :Report result
- Loss on Drying:≤7,0%
Specification Test Results
About this item
:
:342.29648 g/mol
Storage Temperature:Ambient
CAS Number:9050-36-6
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00146679