Fluorescent Brightener 28, powder microbiology stain
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Calcofluor White M2R, Tinopal UNPA-GX
CAS Number:4404-43-7
:
Formula:C₄₀H₄₄N₁₂O₁₀S₂
MDL Number:MFCD00308955
Molecular Weight:916.986 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Synonyms:Calcofluor White M2R; Tinopal UNPA-GX; C.I. 40622
- Form:Yellow Powder
- Storage:Store at Room Temperature (15 - 30 °C)
- Typical Working Concentration:0,05 - 0,20%
Specification Test Results
Frequently Bought Together