Soy hydrolysate enzymatic, powder, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Soy hydrolysate is an enzymatic (papain) digest of soybean meal. It is a plant source peptone. Soy hydrolysate is useful in media where high clarity is not required.
- Total Nitrogen- ≥8,0%
- Nitrogen, AN/TN - 17,0 - 28,0
- Std. Plate Count ≤10 000
- Enterobacteriaceae ≤10 CFU/G
Soluble in 100 mg/ml water.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together