L(+)-Leucine, white powder
: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
:61-90-5
:
:C₆H₁₃NO₂
:MFCD00002617
:131.17 g/mol
:293 °C
:14,05451
:Ambient
- Environmentally Preferable:
- Identity Test:Passes
- FTIR:Conforms to Standard
- Specific Rotation:+14,5 ° to +16,5 °
- Loss on drying:≤0.2%
Frequently Bought Together