XTT sodium salt
Supplier: PanReac AppliChem
Certificates
About this item
Synonyms: 2,3-Bis(2-methoxy-4-nitro-5-sulphophenyl)-2H-tetrazolium-5-carboxanilide, XTT sodium, Sodium 5,5'-(5-(phenylcarbamoyl)-2H-tetrazole-3-ium-2,3-diyl)bis(4-methoxy-2-nitrobenzenesulphonate), sodium 2,3,-bis(2-methoxy-4-nitro-5-sulphophenyl)-5-[(phenylamino)-carbonyl]-2H-tetrazolium inner salt
CAS Number:111072-31-2
:
Formula:C₂₂H₁₇N₇O₁₃S₂Na
MDL Number:MFCD00083517
Molecular Weight:673.52 g/mol
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- λmax.:283 - 287 nm
- E 1 %/1 cm, 285 nm:14,800 - 16,800
Specification Test Results
Frequently Bought Together