Spill containment trays, SW series
Supplier: NEW PIG
Certificates
About this item
Lightweight and durable PE spill trays are ideal for storing several small to medium sized containers of liquids.
- Chemical and rust resistant
- Stackable for easy transportation and storage
- Standard trays with high and durable sidewalls; containers sit deep into the sump, minimising any potential splashes
- Spill trays with grate feature an anti-slip, removable grate that keeps containers away from collected liquids
Caution: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.
Frequently Bought Together