Order Entry
Denmark
ContactUsLinkComponent
HyClone™ Earle's 1X Balanced Salt Solution (EBSS): Solution
HyClone™ Earle's 1X Balanced Salt Solution (EBSS): Solution
Catalog # SH30029.02
CAS Number:  
undefined
HyClone™ Earle's 1X Balanced Salt Solution (EBSS): Solution
Catalog # SH30029.02
CAS Number:  

Specifications

  • Pk:
    500 ml
  • Description:
    HyClone Earle's 1X Balanced Salt Solution
  • Seq:
    02
  • Environmentally Preferable:

Specifications

About this item

Balanced salt solution for cell culture applications.

  • All formulations incorporate a rigorous specification and approval process
  • Available in lot sizes that facilitate scale-up and manufacturing
  • Endotoxin Concentration 1.0 EU/mL
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Caution: Shelf life 24 months. Storage 15° to 3° C