About this item
Temperature gradient plate for determining minimum film forming temperature (MFFT) test. The instrument can be used to define the white point/glass transition temperature of dispersion materials, synthetic resins, enamels and the blocking power and stacking capability of coated papers, foils, prints.
- Ground hard-chrome plated metal platen as to deposit the specimen
- Displays nominal and actual value of hot and cold side
- Electronic controller with digital display, temperature preselecting and overshoot protection
- Integrated membrane drying unit with flow meter
The temperature is controlled through an integrated digital controller with a digital display and measuring-point selector that covers the whole range. To determine the blocking power the specimens on the platen are weighed by defined loads to simulate the stacking capability.
MFFT 10: Includes 10 built-in temperature sensors and hinged transparent cover made of PMMA. Recommended standard cryostat type is C10K20.
MFFT 20: Includes 20 built-in temperature sensors and hinged transparent cover made of stainless steel and safety glass for temperatures up to +250 °C. Recommended standard cryostat type is KWK10.
W×D×H (with closed cover): 350×800×320 mm