Lithium bromide, anhydrous 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7550-35-8
:23303-71-1
Formula:LiBr
MDL Number:MFCD00011077
Boiling Pt:1265 °C (760 mmHg)
Melting Pt:550 °C
Density:3.46 g/cm³
Molecular Weight:86.84 g/mol
Merck Index:13,05548
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥98.5 %
- pH:6 to 12 (4 % in water)
Specification Test Results
Frequently Bought Together