1,3-Di-tert-butylbenzene 99%
:1014-60-4
:
:C₁₄H₂₂
:MFCD00008830
:106…107 °C (18 mmHg)
:10…11 °C
:0.86 g/cm³
:190.33
:84 °C
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4870 to 1.4890 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together