Order Entry
Denmark
ContactUsLinkComponent
Disodium oxalate ACS
Disodium oxalate ACS
Certificates
undefined
Disodium oxalate ACS
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤0.01 %
  • Titration with KMnO4:
    ≥99.5 %
  • Substance darkened by H2SO4:
    Passes test
  • Chloride (Cl):
    ≤0.002 %
  • Sulfate (SO4):
    ≤0.002 %
  • Ammonium (NH4):
    ≤0.002 %
  • Iron (Fe):
    ≤0.001 %
  • Potassium (K):
    ≤0.005 %
  • Heavy metals (as Pb):
    ≤0.002 %
  • Insoluble matter:
    ≤0.005 %
  • Neutrality:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together