2-Methyl-1,3-benzoxazol-5-amine 97%
: 5-Amino-2-methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazol-5-amine
:72745-76-7
:
:C₈H₈N₂O
:MFCD00628970
:148.161 g/mol
:72…74.6 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Off-white to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:70 to 76 °C
- HPLC:≥96.0 %
Frequently Bought Together