3-Benzyloxy-1-propanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Propanol, 3-(benzyloxy)-, 3-phenylmethoxy-1-propanol, Trimethylene glycol monobenzyl ether, 3-(Benzyloxy)-1-propanol, 3-(Benzyloxy)propan-1-ol
CAS Number:4799-68-2
:
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00029659
Boiling Pt:111…114 °C (2 mmHg)
Density:1.049 g/cm³
Molecular Weight:166.22 g/mol
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together