3-Chloropicolinic acid 97%
Catalog # ACRO429912500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:124 to 127 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
Synonyms: 3-Chloropyridine-2-carboxylic acid
:
:157.56 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:57266-69-0
Formula:C₆H₄ClNO₂
MDL Number:MFCD01646325
Melting Pt:124…126 °C