N-(tert-Butoxycarbonyl)-1,4-phenylenediamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (4-Aminophenyl)carbamic Acid tert-Butyl Ester, 4-(tert-Butoxycarbonylamino)aniline, N-BOC-p-phenylenediamine, tert-Butyl (4-aminophenyl)carbamate, 4-(Boc-amino)aniline, N-Boc-1,4-phenylenediamine, tert-Butyl (4-Aminophenyl)carbamate
CAS Number:71026-66-9
:
Formula:C₁₁H₁₆N₂O₂
MDL Number:MFCD00043022
Molecular Weight:208.26 g/mol
Melting Pt:113 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Beige to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:113 to 115 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together