Disposable earplugs, uvex xact-fit detec
Supplier: UVEX
Certificates
About this item
The xact-fit detec is a detectable disposable earplug for use in production areas. The connecting cord makes it practical to wear and limits the risk of process contamination.
- Design adapts perfectly to the shape of the ear reducing pressure on the ear canal, even when worn for long periods
- Stems are marked for left and right ears giving improved wearer comfort and easy, accurate insertion and removal
- Cord and stem reduce the potential for contamination as there is no need to touch the foam earplug before insertion
- Blue colour makes the earplugs easier to identify during the manufacturing process
SNR: 26 dB
EN 352-2:2002
Certifications: EN 352-2:2002
Frequently Bought Together