Viscosity standards, general purpose
Supplier: CANNON INSTRUMENT
Certificates
About this item
These certified standards are useful for calibration and verification procedures.
- It is suitable for all types of viscometers
- Viscosity data quoted at 20 °C
- It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together