Phosphonitrilic chloride trimer 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene
CAS Number:940-71-6
:
Formula:Cl₆N₃P₃
MDL Number:MFCD00006474
Density:1.98 g/cm³
Molecular Weight:347.64 g/mol
Melting Pt:111…115 °C
Storage Temperature:Ambient
UN:3260
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:111 to 115 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together