Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Bz-Arg-OEt.HCl, Nα-Benzoyl-L-arginine Ethyl Ester HCl
CAS Number:2645-08-1
:
Formula:C₁₅H₂₂N₄O₃
MDL Number:MFCD00012579
Molecular Weight:342.82 g/mol
Melting Pt:129…136 °C
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Solubility:(5 % in water) Clear colorless
- TLC:≥99.0 %
- Specific optical rotation:-15.0° to -19.0° (24°C, 589 nm) (c=2, water)
Specification Test Results
Frequently Bought Together