Order Entry
Denmark
ContactUsLinkComponent
CAB-O-SIL® M-5
CAB-O-SIL® M-5
Catalog # ACRO403731500
CAS Number:  
undefined
CAB-O-SIL® M-5
Catalog # ACRO403731500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    150 g
  • Seq:
    3475
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Density:
    ≤60 g/l tamped density
  • Particle size:
    ≤0.02 % 325 Mesh residue
  • Surface area:
    175 to 225 m²/g (BET)
  • pH:
    3.8 to 4.3 (4 % slurry in water)
  • Loss on heating:
    ≤1.5 % (105°C, 2 h)

Specification Test Results

About this item

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
 :
 :60.08 g/mol
Storage Temperature:Ambient
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
Boiling Pt:2230
Melting Pt:1719
Merck Index:14,08493