N-Palmitoyl ceramide (synthetic) 98+%, 99+% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: D-erythro-Sphingosine, N-Palmitoyl-, N-Palmitoyl-D-erythro-sphingosine, C16 Ceramide, N-Hexadecanoyl-D-erythro-sphingosine
CAS Number:24696-26-2
:
Formula:C₃₄H₆₇NO₃
MDL Number:MFCD00871748
Molecular Weight:537.914 g/mol
Melting Pt:94…95 °C
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.50 %
- Enantiomeric excess:≥99.0 %
Specification Test Results
Frequently Bought Together