Order Entry
Denmark
ContactUsLinkComponent
Anti-V5 Tag Rabbit Polyclonal Antibody (CF594)
Anti-V5 Tag Rabbit Polyclonal Antibody (CF594)
Catalog # 20442.
Supplier:  Biotium
Anti-V5 Tag Rabbit Polyclonal Antibody (CF594)
Catalog # 20442.
Supplier:  Biotium

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    V5 tag
  • Antigen symbol:
    V5
  • Clonality:
    Polyclonal
  • Conjugation:
    CF® 594
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
    Store at –20 °C.
  • Concentration:
    1 mg/mL
  • Purification:
    Affinity purified
  • Size:
    100 μL
  • Pk:
    100 µl

Specifications

About this item

Polyclonal anti-V5 (GKPIPNPLLGLDST) reacts with V5 tagged fusion proteins. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus 5 (SV5). The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Tag
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Rabbit
Isotype:
Reactivity: V5 tag