Order Entry
Denmark
ContactUsLinkComponent
Gas lighter, Piezoelectric, SP Bel-Art
Gas lighter, Piezoelectric, SP Bel-Art
Catalog # BELAH150560000
undefined
Gas lighter, Piezoelectric, SP Bel-Art
Catalog # BELAH150560000
Specifications
  • Description:
    Piezolelectric gas lighter
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
For lighting Bunsen burners, gas stoves, propane or oxy-acetylene torches safely.
  • Does not require batteries or fuel refills
  • Comfortable plastic pistol-grip with push-button trigger creates an electrical spark (not a flame) at 20 kv with each squeeze
Length: 265 mm
Weight: 88 g