Order Entry
Denmark
ContactUsLinkComponent
Micrococcal nuclease solution (≥100 U/µl)
Micrococcal nuclease solution (≥100 U/µl)
Catalog # PIER88216
CAS Number:  
undefined
Micrococcal nuclease solution (≥100 U/µl)
Catalog # PIER88216
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Micrococcal Nuclease Solution (≥100 U/µL), 150 µl
  • Environmentally Preferable:
Specifications
About this item
This Micrococcal Nuclease (MNase) is suitable for removing nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experiments.
  • Enzyme digests nucleic acids (DNA and RNA)
  • Effective for double-stranded, single-stranded, circular and linear nucleic acids
  • Active in neutral to alkaline buffer conditions containing divalent calcium ions
  • Highest activity is toward single-stranded nucleic acid substrates with preference for AT or AU rich regions