Dialysis tubing, SnakeSkin™, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
SnakeSkin dialysis tubings with pleated format are made from high performance regenerated-cellulose membrane which has minimal nonspecific binding and a reliable molecular-weight cutoff (MWCO).
- Ready-to-use, pre-opened and pre-wetted
- Greater than 90% protein recovery
Regenerated cellulose is exceptionally stable when exposed to common laboratory solutions such as acidic and basic conditions, or hydrophobic solvents and alcohols. Ideal for dialysing protein samples.
Delivery: Supplied in eight-inch (20 cm) cylinders containing 35 feet (10,7 m) of tubing.
Frequently Bought Together