Order Entry
Denmark
ContactUsLinkComponent
Pierce™ Sealing Tape for 96-Well Microplates
  391-0649
undefined
Pierce™ Sealing Tape for 96-Well Microplates
  391-0649
  • Description:
    Sealing tape, clear acetate with acrylic adhesive, pre-cut sheets, for 96-well microplates
  • Environmentally Preferable:
  • Pk:
    100
Microplate sealing tape consisting of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.
  • Pressure-sensitive, just position the tape over plate and press to seal
  • Each sheet of tape is supplied on its own easy-to-peel backing
  • Durable and inert