Coenzyme A trihydrate
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: CoA trihydrate
CAS Number:85-61-0
:
Formula:C₂₁H₃₆N₇O₁₆P₃S·3H₂O
MDL Number:MFCD00065465
Molecular Weight:821.58 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together