Adipic acid FCC
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid
CAS Number:124-04-9
:
Formula:HOOC(CH₂)₄COOH
MDL Number:MFCD00004420
Boiling Pt:337
Melting Pt:151
Density:1.36 g/cm³ (25 °C)
Molecular Weight:146.14 g/mol
Flash Pt:196 °C (closed cup)
Merck Index:13,00163
EINECS:204-673-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.6% Min.
Specification Test Results
Frequently Bought Together