Diethylene glycol diacrylate ≥75% stabilised, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Diethyleneglycoldiacrylate
CAS Number:4074-88-8
:
Formula:C₁₀H₁₄O₅
MDL Number:MFCD00014939
Boiling Pt:94 °C (0.2 mmHg)
Density:1.118
Molecular Weight:214.22 g/mol
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:223-791-6
UN:2810
ADR:6.1 III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together