Krebs-Ringer solution, HEPES-buffered
Catalog # J67795.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mL
- Description:Krebs-Ringer Solution, HEPES-buffered
- Environmentally Preferable:
- Physical form:Liquid
- Appearance:Colourless
Specifications
About this item
Contains 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, 5,5 mM HEPES, and 1 mM D-glucose
- pH 7,2
- Prepared in 18,2 megohms water
- Filtered through 0,22 µm filter
Physical Form: Liquid
Appearance: Colourless
Standard physiological buffer that is suitable for various calcium imaging experiments. It is also used as a preserving agent.