Krebs-Ringer solution, bicarbonate-buffered
Contains: 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5,5 mM D-glucose.
- pH 7,3
- Filtered through 0,22 micron filter
- Prepared in 18,2 megohms water.
Physical Form: Liquid
Appearance: Colourless
- Environmentally Preferable:
Frequently Bought Together