1-Chlorobutane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: n-Butyl Chloride
CAS Number:109-69-3
:
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009
Boiling Pt:77…78 °C
Melting Pt:–123 °C
Density:0.886
Molecular Weight:92.57 g/mol
Flash Pt:–12 °C (10 °F)
UN:1127
ADR:3 II
Merck Index:13,01559
EINECS:203-696-6
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together