Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl glycolate
tert-Butyl glycolate 94%
tert-Butyl glycolate 94%
tert-Butyl glycolate 94%
H35711.03
:
Thermo Fisher Scientific
:
tert-Butyl glycolate 94%
H35711.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Environmentally Preferable:
:
tert-Butyl 2-hydroxyacetate
:
50595-15-8
:
:
C₆H₁₂O₃
:
MFCD03452739
:
132.159 g/mol
:
GHS07