Cobalt(II,III) oxide (∼3,4 - 4,5% CoO, ∼11.5 - 14.5% MoO) on alumina
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide
CAS Number:1308-06-1
:
Formula:Co₃O₄
MDL Number:MFCD01074445
Molecular Weight:240.8 g/mol
Melting Pt:895
ADR:9 III
Merck Index:13,02454
UN:3077
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together