Anti-NONO Rabbit Polyclonal Antibody
BSBTPB9875
- Antibody type:Primary
- Antigen name:Non-POU domain containing, octamer-binding
- Antigen symbol:NONO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4841
- Antigen synonyms:P54|NRB54|P54NRB|NMT55|PPP1R114
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: NONO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat