Hand cleansing paste, Solopol® natural
Supplier: Deb-STOKO
Certificates
About this item
Solvent-free hand cleanser paste with scrubbing-agent for extra heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust, lubricants. For use in general industrial environments.
- Astopon® natural scrubbing agent gently removes contaminants without damaging the skin
- Contains skin protectant Eucornol® and aloe vera
- Formulation leaves hands feeling smooth and conditioned
- Solvent-free, pH neutral to the skin
Certifications: EU Ecolabel
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together